missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse BAG family molecular chaperone regulator 3 Protein

A DNA sequence encoding the Mus musculus (Mouse) BAG family molecular chaperone regulator 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8071-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16082491

  • £560.00 / 100µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_038891.4
BAG family molecular chaperone regulator 3 Protein
Research Use Only
Bag3
Recombinant Protein
Yeast
SAATQSPMMQ MASGNGASDR DPLPPGWEIK IDPQTGWPFF VDHNSRTTTW NDPRVPPEGP KDTASSANGP SRDGSRLLPI REGHPIYPQL RPGYIPIPVL HEGSENRQPH LFHAYSQPGV QRFRTEAAAA TPQRSQSPLR GGMTEAAQTD KQCGQMPATA TTAAAQPPTA HGPERSQSPA ASDCSSSSSS ASLPSSGRSS LGSHQLPRGY IPIPVIHEQN ITRPAAQPSF HQAQKTHYPA QQGEYQPQQP VYHKIQGDDW EPRPLRAASP FRSPVRGASS REGSPARSGT PVHCPSPIRV HTVVDRPQPM THREPPPVTQ PENKPESKPG PAGPDLPPGH IPIQVIRREA DSKPVSQKSP PPAEKVEVKV SSAPIPCPSP SPAPSAVPSP PKNVAAEQKA APSPAPAEPA APKSGEAETP PKHPGVLKVE AILEKVQGLE QAVDSFEGKK TDKKYLMIEE YLTKELLALD SVDPEGRADV RQARRDGVRK VQTILEKLEQ KAIDVPGQVQ VYELQPSNLE AEQPLQEIMG AVVADKDKKG PENKDPQTES QQLEAKAATP PNPSNPADSA GNLVAP
Greater than 90% as determined by SDS-PAGE.
29810
100 μg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers