missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Angiogenin-4 Protein
A DNA sequence encoding the Mus musculus (Mouse) Angiogenin-4, was expressed in the hosts and tags indicated.
£223.39 - £2410.48
Specifications
Accession Number | NP_808212.2 |
---|---|
Gene ID (Entrez) | 219033 |
Name | Angiogenin-4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048711
|
enQuireBio™
QP9784-YE-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088711
|
enQuireBio™
QP9784-YE-500UG |
500 μg |
£1600.00
500µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058711
|
enQuireBio™
QP9784-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078711
|
enQuireBio™
QP9784-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098711
|
enQuireBio™
QP9784-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068711
|
enQuireBio™
QP9784-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_808212.2 | |
Angiogenin-4 Protein | |
Not determined. | |
Full Length | |
Mouse | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
219033 | |
Research Use Only | |
Ang4 | |
Recombinant Protein | |
Yeast | |
QNERYEKFLR QHYDAKPNGR DDRYCESMMK ERKLTSPCKD VNTFIHGTKK NIRAICGKKG SPYGENFRIS NSPFQITTCT HSGASPRPPC GYRAFKDFRY IVIACEDGWP VHFDESFISP | |
Greater than 90% as determined by SDS-PAGE. |