missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Angiogenin-4 Protein

A DNA sequence encoding the Mus musculus (Mouse) Angiogenin-4, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9784-ye-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16078711

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_808212.2
Angiogenin-4 Protein
Research Use Only
Ang4
Recombinant Protein
Yeast
QNERYEKFLR QHYDAKPNGR DDRYCESMMK ERKLTSPCKD VNTFIHGTKK NIRAICGKKG SPYGENFRIS NSPFQITTCT HSGASPRPPC GYRAFKDFRY IVIACEDGWP VHFDESFISP
Greater than 90% as determined by SDS-PAGE.
219033
200 μg
Not determined.
Full Length
Mouse
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers