missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Mouse Acetyl-coenzyme A synthetase, cytoplasmic Protein
A DNA sequence encoding the Mus musculus (Mouse) Acetyl-coenzyme A synthetase, cytoplasmic, was expressed in the hosts and tags indicated.
£281.57 - £2480.00
Specifications
Accession Number | NP_062785.2 |
---|---|
Gene ID (Entrez) | 60525 |
Name | Acetyl-coenzyme A synthetase, cytoplasmic Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16093111
|
enQuireBio™
QP8182-EC-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16013121
|
enQuireBio™
QP8182-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16003121
|
enQuireBio™
QP8182-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16023121
|
enQuireBio™
QP8182-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16033121
|
enQuireBio™
QP8182-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16043121
|
enQuireBio™
QP8182-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_062785.2 | |
Acetyl-coenzyme A synthetase, cytoplasmic Protein | |
Not determined. | |
Full Length | |
Mouse | |
His | |
Tris-based buffer, 50% glycerol |
60525 | |
Research Use Only | |
Acss2 | |
Recombinant Protein | |
E. coli | |
MGLPEERRKS GSGSRAREET GAEGRVRGWS PPPEVRRSAH VPSLQRYREL HRRSVEEPRE FWGNIAKEFY WKTACPGPFL QYNFDVTKGK IFTEWMKGAT TNICYNVLDR NVHEKKLGDK VAFYWEGNEP GETTKITYRE LLVQVCQFSN VLRKQGIQKG DRVAIYMPMI LELVVAMLAC ARLGALHSIV FAGFSAESLC ERILDSSCCL LITTDAFYRG EKLVNLKELA DESLEKCREK GFPVRCCIVV KHLGRAELGM NDSPSQSPPV KRPCPDVQIC WNEGVDLWWH ELMQQAGDEC EPEWCDAEDP LFILYTSGST GKPKGVVHTI GGYMLYVATT FKYVFDFHPE DVFWCTADIG WITGHSYVTY GPLANGATSV LFEGIPTYPD EGRLWSIVDK YKVTKFYTAP TAIRMLMKFG DDPVTKHSRA SLQVLGTVGE PINPEAWLWY HRVVGSQRCP IVDTFWQTET GGHMLTPLPG ATPMKPGSAS FPFFGVAPAI LNESGEELEG EAEGYLVFKQ PWPGIMRTVY GNHTRFETTY FKKFPGYYVT GDGCRRDQDG YYWITGRIDD MLNVSGHLLS TAEVESALVE HEAVAEAAVV GHPHPVKGEC LYCFVTLCDG HTFSPTLTEE LKKQIREKIG PIATPDYIQN APGLPKTRSG KIMRRVLRKI AQNDHDLGDT STVADPSVIN HLFSHRCLTT Q | |
Greater than 90% as determined by SDS-PAGE. |