missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Mouse Acetyl-coenzyme A synthetase, cytoplasmic Protein

A DNA sequence encoding the Mus musculus (Mouse) Acetyl-coenzyme A synthetase, cytoplasmic, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8182-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16013121

  • £2480.00 / 1mg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_062785.2
Acetyl-coenzyme A synthetase, cytoplasmic Protein
Research Use Only
Acss2
Recombinant Protein
E. coli
MGLPEERRKS GSGSRAREET GAEGRVRGWS PPPEVRRSAH VPSLQRYREL HRRSVEEPRE FWGNIAKEFY WKTACPGPFL QYNFDVTKGK IFTEWMKGAT TNICYNVLDR NVHEKKLGDK VAFYWEGNEP GETTKITYRE LLVQVCQFSN VLRKQGIQKG DRVAIYMPMI LELVVAMLAC ARLGALHSIV FAGFSAESLC ERILDSSCCL LITTDAFYRG EKLVNLKELA DESLEKCREK GFPVRCCIVV KHLGRAELGM NDSPSQSPPV KRPCPDVQIC WNEGVDLWWH ELMQQAGDEC EPEWCDAEDP LFILYTSGST GKPKGVVHTI GGYMLYVATT FKYVFDFHPE DVFWCTADIG WITGHSYVTY GPLANGATSV LFEGIPTYPD EGRLWSIVDK YKVTKFYTAP TAIRMLMKFG DDPVTKHSRA SLQVLGTVGE PINPEAWLWY HRVVGSQRCP IVDTFWQTET GGHMLTPLPG ATPMKPGSAS FPFFGVAPAI LNESGEELEG EAEGYLVFKQ PWPGIMRTVY GNHTRFETTY FKKFPGYYVT GDGCRRDQDG YYWITGRIDD MLNVSGHLLS TAEVESALVE HEAVAEAAVV GHPHPVKGEC LYCFVTLCDG HTFSPTLTEE LKKQIREKIG PIATPDYIQN APGLPKTRSG KIMRRVLRKI AQNDHDLGDT STVADPSVIN HLFSHRCLTT Q
Greater than 90% as determined by SDS-PAGE.
60525
1 mg
Not determined.
Full Length
Mouse
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers