missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human WD repeat-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) WD repeat-containing protein 5, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_060058.1 |
---|---|
Gene ID (Entrez) | 11091 |
Name | WD repeat-containing protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16056540
|
enQuireBio™
QP6884-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16076540
|
enQuireBio™
QP6884-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16066540
|
enQuireBio™
QP6884-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16086540
|
enQuireBio™
QP6884-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16096540
|
enQuireBio™
QP6884-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16006550
|
enQuireBio™
QP6884-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Spezifikation
NP_060058.1 | |
WD repeat-containing protein 1 | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
11091 | |
Research Use Only | |
WDR5 | |
Recombinant Protein | |
E. coli | |
ATEEKKPETE AARAQPTPSS SATQSKPTPV KPNYALKFTL AGHTKAVSSV KFSPNGEWLA SSSADKLIKI WGAYDGKFEK TISGHKLGIS DVAWSSDSNL LVSASDDKTL KIWDVSSGKC LKTLKGHSNY VFCCNFNPQS NLIVSGSFDE SVRIWDVKTG KCLKTLPAHS DPVSAVHFNR DGSLIVSSSY DGLCRIWDTA SGQCLKTLID DDNPPVSFVK FSPNGKYILA ATLDNTLKLW DYSKGKCLKT YTGHKNEKYC IFANFSVTGG KWIVSGSEDN LVYIWNLQTK EIVQKLQGHT DVVISTACHP TENIIASAAL ENDKTIKLWK SDC | |
Greater than 90% as determined by SDS-PAGE. |