missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human WD repeat-containing protein 1

A DNA sequence encoding the Homo sapiens (Human) WD repeat-containing protein 5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6884-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16076540

  • £1735.00 / 1mg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_060058.1
WD repeat-containing protein 1
Research Use Only
WDR5
Recombinant Protein
E. coli
ATEEKKPETE AARAQPTPSS SATQSKPTPV KPNYALKFTL AGHTKAVSSV KFSPNGEWLA SSSADKLIKI WGAYDGKFEK TISGHKLGIS DVAWSSDSNL LVSASDDKTL KIWDVSSGKC LKTLKGHSNY VFCCNFNPQS NLIVSGSFDE SVRIWDVKTG KCLKTLPAHS DPVSAVHFNR DGSLIVSSSY DGLCRIWDTA SGQCLKTLID DDNPPVSFVK FSPNGKYILA ATLDNTLKLW DYSKGKCLKT YTGHKNEKYC IFANFSVTGG KWIVSGSEDN LVYIWNLQTK EIVQKLQGHT DVVISTACHP TENIIASAAL ENDKTIKLWK SDC
Greater than 90% as determined by SDS-PAGE.
11091
1 mg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers