missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transaldolase Protein
A DNA sequence encoding the Homo sapiens (Human) Transaldolase, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_006746.1 |
---|---|
Gene ID (Entrez) | 6888 |
Name | Transaldolase Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16084801
|
enQuireBio™
QP8473-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16004811
|
enQuireBio™
QP8473-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16094801
|
enQuireBio™
QP8473-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014811
|
enQuireBio™
QP8473-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024811
|
enQuireBio™
QP8473-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034811
|
enQuireBio™
QP8473-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_006746.1 | |
Transaldolase Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
6888 | |
Research Use Only | |
TAL | |
Recombinant Protein | |
E. coli | |
QRMESALDQL KQFTTVVADT GDFHAIDEYK PQDATTNPSL ILAAAQMPAY QELVEEAIAY GRKLGGSQED QIKNAIDKLF VLFGAEILKK IPGRVSTEVD ARLSFDKDAM VARARRLIEL YKEAGISKDR ILIKLSSTWE GIQAGKELEE QHGIHCNMTL LFSFAMS-QA VACAEAGVTL ISPFVGRILD WHVANTDKKS YEPLEDPGVK SVTKIYNYYK KFSYKTIVMG ASFRNTGEIK ALAGCDFLTI SPKLLGELLQ DNAKLVPVLS AKAAQASDLE KIHLDEKSFR WLHNEDQMAV EKLSDGIRKF AADAVKLERM LTERMFNAEN GK | |
Greater than 90% as determined by SDS-PAGE. |