missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Transaldolase Protein
A DNA sequence encoding the Homo sapiens (Human) Transaldolase, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8473-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_006746.1 | |
Transaldolase Protein | |
Research Use Only | |
TAL | |
Recombinant Protein | |
E. coli | |
QRMESALDQL KQFTTVVADT GDFHAIDEYK PQDATTNPSL ILAAAQMPAY QELVEEAIAY GRKLGGSQED QIKNAIDKLF VLFGAEILKK IPGRVSTEVD ARLSFDKDAM VARARRLIEL YKEAGISKDR ILIKLSSTWE GIQAGKELEE QHGIHCNMTL LFSFAMS-QA VACAEAGVTL ISPFVGRILD WHVANTDKKS YEPLEDPGVK SVTKIYNYYK KFSYKTIVMG ASFRNTGEIK ALAGCDFLTI SPKLLGELLQ DNAKLVPVLS AKAAQASDLE KIHLDEKSFR WLHNEDQMAV EKLSDGIRKF AADAVKLERM LTERMFNAEN GK | |
Greater than 90% as determined by SDS-PAGE. |
6888 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |