missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGF-beta 1 / TGFB1 Protein
A DNA sequence encoding the Homo sapiens (Human) TGF-beta 1 / TGFB1, was expressed in the hosts and tags indicated.
£145.46 - £1688.38
Specifications
Accession Number | NP_000651.3 |
---|---|
Gene ID (Entrez) | 7040 |
Name | TGF-beta 1 / TGFB1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16075331
|
enQuireBio™
QP8568-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16015341
|
enQuireBio™
QP8568-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16065331
|
enQuireBio™
QP8568-EC-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16095331
|
enQuireBio™
QP8568-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16005341
|
enQuireBio™
QP8568-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16085331
|
enQuireBio™
QP8568-EC-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000651.3 | |
TGF-beta 1 / TGFB1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7040 | |
Research Use Only | |
TGFB1 | |
Recombinant Protein | |
E. coli | |
DTNYCFSSTE KNCCVRQLYI DFRKDLGWKW IHEPKGYHAN FCLGPCPYIW SLDTQYSKVL ALYNQHNPGA SAAPCCVPQA LEPLPIVYYV GRKPKVEQLS NMIVRSCKCS | |
Greater than 90% as determined by SDS-PAGE. |