missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human TGF-beta 1 / TGFB1 Protein
A DNA sequence encoding the Homo sapiens (Human) TGF-beta 1 / TGFB1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP8568-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_000651.3 | |
TGF-beta 1 / TGFB1 Protein | |
Research Use Only | |
TGFB1 | |
Recombinant Protein | |
E. coli | |
DTNYCFSSTE KNCCVRQLYI DFRKDLGWKW IHEPKGYHAN FCLGPCPYIW SLDTQYSKVL ALYNQHNPGA SAAPCCVPQA LEPLPIVYYV GRKPKVEQLS NMIVRSCKCS | |
Greater than 90% as determined by SDS-PAGE. |
7040 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |