missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human TGF-beta 1 / TGFB1 Protein

A DNA sequence encoding the Homo sapiens (Human) TGF-beta 1 / TGFB1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP8568-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16005341

  • £1075.37 / 500µg

Estimated Shipment
03-11-2020

Please to purchase this item. Need a web account? Register with us today!


Specifications

Specifications

NP_000651.3
TGF-beta 1 / TGFB1 Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
7040
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TGFB1
Recombinant Protein
E. coli
DTNYCFSSTE KNCCVRQLYI DFRKDLGWKW IHEPKGYHAN FCLGPCPYIW SLDTQYSKVL ALYNQHNPGA SAAPCCVPQA LEPLPIVYYV GRKPKVEQLS NMIVRSCKCS
Videos
Documents