missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Surfactant-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) Surfactant-associated protein 3, was expressed in the hosts and tags indicated.
£439.00 - £1230.00
Specifications
Accession Number | NP_001094811.1 |
---|---|
Gene ID (Entrez) | 253970 |
Name | Surfactant-associated protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16095040
|
enQuireBio™
QP6680-MA-20UG |
20 μg |
£439.00
20µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16085040
|
enQuireBio™
QP6680-MA-100UG |
100 μg |
£1230.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16005050
|
enQuireBio™
QP6680-MA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001094811.1 | |
Surfactant-associated protein 1 | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
253970 | |
Research Use Only | |
SFTA3 | |
Recombinant Protein | |
Mammal | |
MRAGFSDFQL IRDQVLFLQD QAQRLTEWLQ LSGFENPVSE STTLCLRERE KRIPTCVAVC VPSPGTVHTA LLHPTTLSQS RSSSEAKMLI IHTA | |
Greater than 90% as determined by SDS-PAGE. |