missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Surfactant-associated protein 1

A DNA sequence encoding the Homo sapiens (Human) Surfactant-associated protein 3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6680-ma-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16085040

  • £1230.00 / 100µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001094811.1
Surfactant-associated protein 1
Research Use Only
SFTA3
Recombinant Protein
Mammal
MRAGFSDFQL IRDQVLFLQD QAQRLTEWLQ LSGFENPVSE STTLCLRERE KRIPTCVAVC VPSPGTVHTA LLHPTTLSQS RSSSEAKMLI IHTA
Greater than 90% as determined by SDS-PAGE.
253970
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers