missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SFTPD / SP-D / SFTP4 Protein
A DNA sequence encoding the Homo sapiens (Human) SFTPD / SP-D / SFTP4, was expressed in the hosts and tags indicated.
£223.39 - £2410.48
Specifications
Accession Number | NP_003010.4 |
---|---|
Gene ID (Entrez) | 6441 |
Name | SFTPD / SP-D / SFTP4 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16048241
|
enQuireBio™
QP9395-YE-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16088241
|
enQuireBio™
QP9395-YE-500UG |
500 μg |
£1600.00
500µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16058241
|
enQuireBio™
QP9395-YE-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16078241
|
enQuireBio™
QP9395-YE-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16098241
|
enQuireBio™
QP9395-YE-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16068241
|
enQuireBio™
QP9395-YE-1MG |
1 mg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_003010.4 | |
SFTPD / SP-D / SFTP4 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
6441 | |
Research Use Only | |
SFTPD | |
Recombinant Protein | |
Yeast | |
AEMKTYSHRT MPSACTLVMC SSVESGLPGR DGRDGREGPR GEKGDPGLPG AAGQAGMPGQ AGPVGPKGDN GSVGEPGPKG DTGPSGPPGP PGVPGPAGRE GPLGKQGNIG PQGKPGPKGE AGPKGEVGAP GMQGSAGARG LAGPKGERGV PGERGVPGNT GAAGSAGAMG PQGSPGARGP PGLKGDKGIP GDKGAKGESG LPDVASLRQQ VEALQGQVQH LQAAFSQYKK VELFPNGQSV GEKIFKTAGF VKPFTEAQLL CTQAGGQLAS PRSAAENAAL QQLVVAKNEA AFLSMTDSKT EGKFTYPTGE SLVYSNWAPG EPNDDGGSED CVEIF | |
Greater than 90% as determined by SDS-PAGE. |