missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SFTPD / SP-D / SFTP4 Protein
A DNA sequence encoding the Homo sapiens (Human) SFTPD / SP-D / SFTP4, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP9395-ye-200ug
Additional Details : Weight : 0.01000kg
Specifications
NP_003010.4 | |
SFTPD / SP-D / SFTP4 Protein | |
Research Use Only | |
SFTPD | |
Recombinant Protein | |
Yeast | |
AEMKTYSHRT MPSACTLVMC SSVESGLPGR DGRDGREGPR GEKGDPGLPG AAGQAGMPGQ AGPVGPKGDN GSVGEPGPKG DTGPSGPPGP PGVPGPAGRE GPLGKQGNIG PQGKPGPKGE AGPKGEVGAP GMQGSAGARG LAGPKGERGV PGERGVPGNT GAAGSAGAMG PQGSPGARGP PGLKGDKGIP GDKGAKGESG LPDVASLRQQ VEALQGQVQH LQAAFSQYKK VELFPNGQSV GEKIFKTAGF VKPFTEAQLL CTQAGGQLAS PRSAAENAAL QQLVVAKNEA AFLSMTDSKT EGKFTYPTGE SLVYSNWAPG EPNDDGGSED CVEIF | |
Greater than 90% as determined by SDS-PAGE. |
6441 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |