missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SerpinB1 / ELANH2 Protein
A DNA sequence encoding the Homo sapiens (Human) SerpinB1 / ELANH2, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_109591.1 |
---|---|
Gene ID (Entrez) | 1992 |
Name | SerpinB1 / ELANH2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16084950
|
enQuireBio™
QP6668-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16004960
|
enQuireBio™
QP6668-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16094950
|
enQuireBio™
QP6668-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014960
|
enQuireBio™
QP6668-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024960
|
enQuireBio™
QP6668-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034960
|
enQuireBio™
QP6668-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_109591.1 | |
SerpinB1 / ELANH2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
1992 | |
Research Use Only | |
SerpinB1 | |
Recombinant Protein | |
E. coli | |
MEQLSSANTR FALDLFLALS ENNPAGNIFI SPFSISSAMA MVFLGTRGNT AAQLSKTFHF NTVEEVHSRF QSLNADINKR GASYILKLAN RLYGEKTYNF LPEFLVSTQK TYGADLASVD FQHASEDARK TINQWVKGQT EGKIPELLAS GMVDNMTKLV LVNAIYFKGN WKDKFMKEAT TNAPFRLNKK DRKTVKMMYQ KKKFAYGYIE DLKCRVLELP YQGEELSMVI LLPDDIEDES TGLKKIEEQL TLEKLHEWTK PENLDFIEVN VSLPRFKLEE SYTLNSDLAR LGVQDLFNSS KADLSGMSGA RDIFISKIVH KSFVEVNEEG TEAAAATAGI ATFCMLMPEE NFTADHPFLF FIRHNSSGSI LFLGRFSSP | |
Greater than 90% as determined by SDS-PAGE. |