missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human SerpinB1 / ELANH2 Protein
A DNA sequence encoding the Homo sapiens (Human) SerpinB1 / ELANH2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6668-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_109591.1 | |
SerpinB1 / ELANH2 Protein | |
Research Use Only | |
SerpinB1 | |
Recombinant Protein | |
E. coli | |
MEQLSSANTR FALDLFLALS ENNPAGNIFI SPFSISSAMA MVFLGTRGNT AAQLSKTFHF NTVEEVHSRF QSLNADINKR GASYILKLAN RLYGEKTYNF LPEFLVSTQK TYGADLASVD FQHASEDARK TINQWVKGQT EGKIPELLAS GMVDNMTKLV LVNAIYFKGN WKDKFMKEAT TNAPFRLNKK DRKTVKMMYQ KKKFAYGYIE DLKCRVLELP YQGEELSMVI LLPDDIEDES TGLKKIEEQL TLEKLHEWTK PENLDFIEVN VSLPRFKLEE SYTLNSDLAR LGVQDLFNSS KADLSGMSGA RDIFISKIVH KSFVEVNEEG TEAAAATAGI ATFCMLMPEE NFTADHPFLF FIRHNSSGSI LFLGRFSSP | |
Greater than 90% as determined by SDS-PAGE. |
1992 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |