missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100A7 Protein
A cDNA sequence encoding the S100A7 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | S100A7 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15973439
|
enQuireBio™
QP13398-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15963439
|
enQuireBio™
QP13398-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953439
|
enQuireBio™
QP13398-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
S100A7 Protein | |
Not Determined. Testing recommended prior to use in cell culture. | |
Human | |
His | |
S100A7 protein at 0.5 mg/ml in 20mM Tris-HCl, pH 8.0, 0.1M NaCl, 1mM DTT and 30% Glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
HIS-TAG-SSGLVPRGSH MSNTQAERSI IGMIDMFHKY TRRDDKIDKP SLLTMMKENFPNFLSACDKK GTNYLADVFE KKDKNEDKKI DFSEFLSLLG DIATDYHKQS HGAAPCSGGS Q | |
Greater than 95.0% as determined by SDS-PAGE. |