missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human S100A7 Protein

A cDNA sequence encoding the S100A7 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13398-20ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15963439

  • £155.00 / 20µg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

S100A7 Protein
Research Use Only
Recombinant Protein
E. coli
HIS-TAG-SSGLVPRGSH MSNTQAERSI IGMIDMFHKY TRRDDKIDKP SLLTMMKENFPNFLSACDKK GTNYLADVFE KKDKNEDKKI DFSEFLSLLG DIATDYHKQS HGAAPCSGGS Q
Greater than 95.0% as determined by SDS-PAGE.
20 μg
Not Determined. Testing recommended prior to use in cell culture.
Human
His
S100A7 protein at 0.5 mg/ml in 20mM Tris-HCl, pH 8.0, 0.1M NaCl, 1mM DTT and 30% Glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers