missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human S100A7 Protein
A cDNA sequence encoding the S100A7 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13398-1mg
Additional Details : Weight : 0.01000kg
Specifications
S100A7 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
HIS-TAG-SSGLVPRGSH MSNTQAERSI IGMIDMFHKY TRRDDKIDKP SLLTMMKENFPNFLSACDKK GTNYLADVFE KKDKNEDKKI DFSEFLSLLG DIATDYHKQS HGAAPCSGGS Q | |
Greater than 95.0% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture. | |
Human | |
His | |
S100A7 protein at 0.5 mg/ml in 20mM Tris-HCl, pH 8.0, 0.1M NaCl, 1mM DTT and 30% Glycerol. |