missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RNASE3 Protein
A cDNA sequence encoding the RNASE3 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | RNASE3 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15943209
|
enQuireBio™
QP13323-HIS-2UG |
2 μg |
£99.15
2µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15953209
|
enQuireBio™
QP13323-HIS-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15923209
|
enQuireBio™
QP13323-HIS-10UG |
10 μg |
£155.00
10µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15933209
|
enQuireBio™
QP13323-HIS-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15963209
|
enQuireBio™
QP13323-HIS-EC-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
RNASE3 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RNASE3 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
ADPRPPQFTR AQWFAIQHIS LNPPRCTIAM RAINNYRWRC KNQNTFLRTT FANVVNVCGN QSIRCPHNRT LNNCHRSRFR VPLLHCDLIN PGAQNISNCR YADRPGRRFY VVACDNRDPR DSPRYPVVPV HLDTTIHHHH HH | |
Greater than 95.0% as determined by SDS-PAGE. |