missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human RNASE3 Protein
A cDNA sequence encoding the RNASE3 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP13323-HIS-5ug
Additional Details : Weight : 0.01000kg
Specifications
RNASE3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
ADPRPPQFTR AQWFAIQHIS LNPPRCTIAM RAINNYRWRC KNQNTFLRTT FANVVNVCGN QSIRCPHNRT LNNCHRSRFR VPLLHCDLIN PGAQNISNCR YADRPGRRFY VVACDNRDPR DSPRYPVVPV HLDTTIHHHH HH | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
RNASE3 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |