missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human RNASE3 Protein

A cDNA sequence encoding the RNASE3 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP13323-HIS-10ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15923209

  • £155.00 / 10µg
Estimated Shipment: 14-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

RNASE3 Protein
Research Use Only
Recombinant Protein
E. coli
ADPRPPQFTR AQWFAIQHIS LNPPRCTIAM RAINNYRWRC KNQNTFLRTT FANVVNVCGN QSIRCPHNRT LNNCHRSRFR VPLLHCDLIN PGAQNISNCR YADRPGRRFY VVACDNRDPR DSPRYPVVPV HLDTTIHHHH HH
Greater than 95.0% as determined by SDS-PAGE.
10 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
RNASE3 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers