missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Rhomboid-related protein 1
A DNA sequence encoding the Homo sapiens (Human) Rhomboid-related protein 2, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_060291.2 |
---|---|
Gene ID (Entrez) | 54933 |
Name | Rhomboid-related protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16084500
|
enQuireBio™
QP6607-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16004510
|
enQuireBio™
QP6607-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16094500
|
enQuireBio™
QP6607-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16014510
|
enQuireBio™
QP6607-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024510
|
enQuireBio™
QP6607-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034510
|
enQuireBio™
QP6607-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_060291.2 | |
Rhomboid-related protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
54933 | |
Research Use Only | |
RHBDL2 | |
Recombinant Protein | |
E. coli | |
MNLNMGREMK EELEEEEKMR EDGGGKDRAK SKKVHRIVSK WMLPEKSRGT YLERANCFPP PVFIISISLA ELAVFIYYAV WKPQKQWITL DTGILESPFI YSPEKREEAW RFISYMLVHA G | |
Greater than 90% as determined by SDS-PAGE. |