missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Rhomboid-related protein 1

A DNA sequence encoding the Homo sapiens (Human) Rhomboid-related protein 2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6607-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16004510

  • £1735.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_060291.2
Rhomboid-related protein 1
Research Use Only
RHBDL2
Recombinant Protein
E. coli
MNLNMGREMK EELEEEEKMR EDGGGKDRAK SKKVHRIVSK WMLPEKSRGT YLERANCFPP PVFIISISLA ELAVFIYYAV WKPQKQWITL DTGILESPFI YSPEKREEAW RFISYMLVHA G
Greater than 90% as determined by SDS-PAGE.
54933
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers