missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Retinol-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) Retinol-binding protein 3, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_002891.1 |
---|---|
Gene ID (Entrez) | 5949 |
Name | Retinol-binding protein 1 |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16004370
|
enQuireBio™
QP6589-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16024370
|
enQuireBio™
QP6589-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16014370
|
enQuireBio™
QP6589-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034370
|
enQuireBio™
QP6589-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044370
|
enQuireBio™
QP6589-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16054370
|
enQuireBio™
QP6589-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002891.1 | |
Retinol-binding protein 1 | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
5949 | |
Research Use Only | |
RBP3 | |
Recombinant Protein | |
E. coli | |
HQSLGALVEG TGHLLEAHYA RPEVVGQTSA LLRAKLAQGA YRTAVDLESL ASQLTADLQE VSGDHRLLVF HSPGELVVEE APPPPPAVPS PEELTYLIEA LFKTEVLPGQ LGYLRFDAMA ELETVKAVGP QLVRLVWQQL VDTAALVIDL RYNPGSYSTA IPLLCSYFFE AEPRQHLYSV FDRATSKVTE VWTLPQVAGQ RYGSHKDLYI LMSHTSGSAA EAFAHTMQDL QRATVIGEPT AGGALSVGIY QVGSSPLYAS MPTQMAMSAT TGKAWDLAGV EPDITVPMSE ALSIAQDIVA LRAKVPTVLQ TAGKLVADNY ASAELGAKMA TKLSGLQSRY SRVTSEVALA EILGADLQML SGDPHLKAAH IPENAKDRIP GIVPMQIPSP EVFEELIKFS FHTNVLEDNI GYLRFDMFGD GELLTQVSRL LVEHIWKKIM HTDAMIIDMR FNIGGPTSSI PILCSYFFDE GPPVLLDKIY SRPDDSVSEL WTHAQVVGER YGSKKSMVIL TSSVTAGTAE EFTYIMKRLG RALVIGEVTS GGCQPPQTYH VDDTNLYLTI PTARSVGASD GSSWEGVGVT PHVVVPAEEA LARAKEMLQ | |
Greater than 90% as determined by SDS-PAGE. |