missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Retinol-binding protein 1
A DNA sequence encoding the Homo sapiens (Human) Retinol-binding protein 3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6589-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002891.1 | |
Retinol-binding protein 1 | |
Research Use Only | |
RBP3 | |
Recombinant Protein | |
E. coli | |
HQSLGALVEG TGHLLEAHYA RPEVVGQTSA LLRAKLAQGA YRTAVDLESL ASQLTADLQE VSGDHRLLVF HSPGELVVEE APPPPPAVPS PEELTYLIEA LFKTEVLPGQ LGYLRFDAMA ELETVKAVGP QLVRLVWQQL VDTAALVIDL RYNPGSYSTA IPLLCSYFFE AEPRQHLYSV FDRATSKVTE VWTLPQVAGQ RYGSHKDLYI LMSHTSGSAA EAFAHTMQDL QRATVIGEPT AGGALSVGIY QVGSSPLYAS MPTQMAMSAT TGKAWDLAGV EPDITVPMSE ALSIAQDIVA LRAKVPTVLQ TAGKLVADNY ASAELGAKMA TKLSGLQSRY SRVTSEVALA EILGADLQML SGDPHLKAAH IPENAKDRIP GIVPMQIPSP EVFEELIKFS FHTNVLEDNI GYLRFDMFGD GELLTQVSRL LVEHIWKKIM HTDAMIIDMR FNIGGPTSSI PILCSYFFDE GPPVLLDKIY SRPDDSVSEL WTHAQVVGER YGSKKSMVIL TSSVTAGTAE EFTYIMKRLG RALVIGEVTS GGCQPPQTYH VDDTNLYLTI PTARSVGASD GSSWEGVGVT PHVVVPAEEA LARAKEMLQ | |
Greater than 90% as determined by SDS-PAGE. |
5949 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |