missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PIN1 Protein
A cDNA sequence encoding the PIN1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £3085.00
Specifications
Gene ID (Entrez) | 5300 |
---|---|
Name | PIN1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Gene Symbol | PIN1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15965268
|
enQuireBio™
QP10820-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15955268
|
enQuireBio™
QP10820-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15945268
|
enQuireBio™
QP10820-1MG |
1 mg |
£3085.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
5300 | |
Research Use Only | |
PIN1 | |
Recombinant Protein | |
E. coli | |
MADEEKLPPG WEKRMSRSSG RVYYFNHITN ASQWERPSGN SSSGGKNGQG EPARVRCSHL LVKHSQSRRP SSWRQEKITR TKEEALELIN GYIQKIKSGE EDFESLASQF SDCSSAKARG DLGAFSRGQM QKPFEDASFA LRTGEMSGPV FTDSGIHIIL RTE | |
Greater than 95.0% as determined by SDS-PAGE. |
PIN1 Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 330 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-HCl pH8.0 using chymotrypsin. | |
Human | |
Untagged | |
The PIN1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 7.5) 0.1M NaCl, 5mM DTT and 20% Glycerol. |