missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PIN1 Protein

A cDNA sequence encoding the PIN1 was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10820-20ug

 View more versions of this product

Product Code. 15955268

This item is not returnable. View return policy

Description

Description

Specifications

Specifications

5300
20 μg
< 1.0 EU per ug protein as determined by the LAL method.
Specific activity is > 330 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-HCl pH8.0 using chymotrypsin.
Human
Untagged
The PIN1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 7.5) 0.1M NaCl, 5mM DTT and 20% Glycerol.
PIN1 Protein
Research Use Only
PIN1
Recombinant Protein
E. coli
MADEEKLPPG WEKRMSRSSG RVYYFNHITN ASQWERPSGN SSSGGKNGQG EPARVRCSHL LVKHSQSRRP SSWRQEKITR TKEEALELIN GYIQKIKSGE EDFESLASQF SDCSSAKARG DLGAFSRGQM QKPFEDASFA LRTGEMSGPV FTDSGIHIIL RTE
Greater than 95.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers