missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PIN1 Protein
A cDNA sequence encoding the PIN1 was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10820-1mg
Additional Details : Weight : 0.01000kg
Specifications
5300 | |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is > 330 nmoles/min/mg, and is defined as the amount of enzyme that cleaves 1umole of suc-AAFP-pNA per minute at 25C in Tris-HCl pH8.0 using chymotrypsin. | |
Human | |
Untagged | |
The PIN1 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 7.5) 0.1M NaCl, 5mM DTT and 20% Glycerol. |
PIN1 Protein | |
Research Use Only | |
PIN1 | |
Recombinant Protein | |
E. coli | |
MADEEKLPPG WEKRMSRSSG RVYYFNHITN ASQWERPSGN SSSGGKNGQG EPARVRCSHL LVKHSQSRRP SSWRQEKITR TKEEALELIN GYIQKIKSGE EDFESLASQF SDCSSAKARG DLGAFSRGQM QKPFEDASFA LRTGEMSGPV FTDSGIHIIL RTE | |
Greater than 95.0% as determined by SDS-PAGE. |