missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDGF CC Protein
A cDNA sequence encoding the PDGF CC was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | PDGF CC Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
Biological Activity | The ED50, as measured in a proliferation assay using mouse NR6R-3T3 cells, is less than 350ng/ml. |
Product Type | Recombinant Protein |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945228
|
enQuireBio™
QP10806-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935228
|
enQuireBio™
QP10806-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925228
|
enQuireBio™
QP10806-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 20-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
PDGF CC Protein | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MHHHHHHVVD LNLLTEEVRL YSCTPRNFSV SIREELKRTD TIFWPGCLLV KRCGGNCACC LHNCNECQCV PSKVTKKYHE VLQLRPKTGV RGLHKSLTDV ALEHHEECDC VCRGSTGG | |
Greater than 97.0% as determined by SDS-PAGE. |
Research Use Only | |
The ED50, as measured in a proliferation assay using mouse NR6R-3T3 cells, is less than 350ng/ml. | |
Human | |
His | |
Lyophilized from a 0.2microm filtered solution in Acetonitrile and TFA. |