missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human PDGF CC Protein
A cDNA sequence encoding the PDGF CC was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10806-20ug
Additional Details : Weight : 0.01000kg
Specifications
PDGF CC Protein | |
Research Use Only | |
The ED50, as measured in a proliferation assay using mouse NR6R-3T3 cells, is less than 350ng/ml. | |
Human | |
His | |
Lyophilized from a 0.2microm filtered solution in Acetonitrile and TFA. |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MHHHHHHVVD LNLLTEEVRL YSCTPRNFSV SIREELKRTD TIFWPGCLLV KRCGGNCACC LHNCNECQCV PSKVTKKYHE VLQLRPKTGV RGLHKSLTDV ALEHHEECDC VCRGSTGG | |
Greater than 97.0% as determined by SDS-PAGE. |