missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human PDGF CC Protein

A cDNA sequence encoding the PDGF CC was constructed and used to recombinantly synthesize the protein.

Brand:  enQuireBio™ QP10806-5ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15945228

  • £99.15 / 5µg
Estimated Shipment: 30-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

PDGF CC Protein
Research Use Only
The ED50, as measured in a proliferation assay using mouse NR6R-3T3 cells, is less than 350ng/ml.
Human
His
Lyophilized from a 0.2microm filtered solution in Acetonitrile and TFA.
5 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
MHHHHHHVVD LNLLTEEVRL YSCTPRNFSV SIREELKRTD TIFWPGCLLV KRCGGNCACC LHNCNECQCV PSKVTKKYHE VLQLRPKTGV RGLHKSLTDV ALEHHEECDC VCRGSTGG
Greater than 97.0% as determined by SDS-PAGE.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers