missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Parathyroid hormone-related Protein
A DNA sequence encoding the Homo sapiens (Human) Parathyroid hormone-related protein, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_002811.1 |
---|---|
Gene ID (Entrez) | 5744 |
Name | Parathyroid hormone-related Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16094100
|
enQuireBio™
QP6553-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16014110
|
enQuireBio™
QP6553-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16004110
|
enQuireBio™
QP6553-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16024110
|
enQuireBio™
QP6553-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16034110
|
enQuireBio™
QP6553-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16044110
|
enQuireBio™
QP6553-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002811.1 | |
Parathyroid hormone-related Protein | |
Not determined. | |
Full length of Isoform 2 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
5744 | |
Research Use Only | |
PTHLH | |
Recombinant Protein | |
E. coli | |
AVSEHQLLHD KGKSIQDLRR RFFLHHLIAE IHTAEIRATS EVSPNSKPSP NTKNHPVRFG SDDEGRYLTQ ETNKVETYKE QPLKTPGKKK KGKPGKRKEQ EKKKRRTRSA WLDSGVTGSG LEGDHLSDTS TTSLELDSR | |
Greater than 90% as determined by SDS-PAGE. |