missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Parathyroid hormone-related Protein
A DNA sequence encoding the Homo sapiens (Human) Parathyroid hormone-related protein, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6553-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_002811.1 | |
Parathyroid hormone-related Protein | |
Research Use Only | |
PTHLH | |
Recombinant Protein | |
E. coli | |
AVSEHQLLHD KGKSIQDLRR RFFLHHLIAE IHTAEIRATS EVSPNSKPSP NTKNHPVRFG SDDEGRYLTQ ETNKVETYKE QPLKTPGKKK KGKPGKRKEQ EKKKRRTRSA WLDSGVTGSG LEGDHLSDTS TTSLELDSR | |
Greater than 90% as determined by SDS-PAGE. |
5744 | |
10 μg | |
Not determined. | |
Full length of Isoform 2 | |
Human | |
His | |
Tris-based buffer, 50% glycerol |