missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Oligodendrocyte transcription factor 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Oligodendrocyte transcription factor 2, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_005797.1 |
---|---|
Gene ID (Entrez) | 10215 |
Name | Oligodendrocyte transcription factor 2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16029740
|
enQuireBio™
QP7608-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16049740
|
enQuireBio™
QP7608-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 12-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16039740
|
enQuireBio™
QP7608-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16059740
|
enQuireBio™
QP7608-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069740
|
enQuireBio™
QP7608-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079740
|
enQuireBio™
QP7608-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005797.1 | |
Oligodendrocyte transcription factor 2 Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
10215 | |
Research Use Only | |
OLIG2 | |
Recombinant Protein | |
E. coli | |
MDSDASLVSS RPSSPEPDDL FLPARSKGSS GSAFTGGTVS SSTPSDCPPE LSAELRGAMG SAGAHPGDKL GGSGFKSSSS STSSSTSSAA ASSTKKDKKQ MTEPELQQLR LKINSRERKR MHDLNIAMDG LREVMPYAHG PSVRKLSKIA TLLLARNYIL MLTNSLEEMK RLVSEIYGGH HAGFHPSACG GLAHSAPLPA ATAHPAAAAH AAHHPAVHHP ILPPAAAAAA AAAAAAAVSS ASLPGSGLPS VGSIRPPHGL LKSPSAAAAA PLGGGGGGSG ASGGFQHWGG MPCPCSMCQV PPPHHHVSAM GAGSLPRLTS DAK | |
Greater than 90% as determined by SDS-PAGE. |