missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Oligodendrocyte transcription factor 2 Protein
A DNA sequence encoding the Homo sapiens (Human) Oligodendrocyte transcription factor 2, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7608-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005797.1 | |
Oligodendrocyte transcription factor 2 Protein | |
Research Use Only | |
OLIG2 | |
Recombinant Protein | |
E. coli | |
MDSDASLVSS RPSSPEPDDL FLPARSKGSS GSAFTGGTVS SSTPSDCPPE LSAELRGAMG SAGAHPGDKL GGSGFKSSSS STSSSTSSAA ASSTKKDKKQ MTEPELQQLR LKINSRERKR MHDLNIAMDG LREVMPYAHG PSVRKLSKIA TLLLARNYIL MLTNSLEEMK RLVSEIYGGH HAGFHPSACG GLAHSAPLPA ATAHPAAAAH AAHHPAVHHP ILPPAAAAAA AAAAAAAVSS ASLPGSGLPS VGSIRPPHGL LKSPSAAAAA PLGGGGGGSG ASGGFQHWGG MPCPCSMCQV PPPHHHVSAM GAGSLPRLTS DAK | |
Greater than 90% as determined by SDS-PAGE. |
10215 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |