missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MED1 Protein
A DNA sequence encoding the Homo sapiens (Human) MED1, was expressed in the hosts and tags indicated.
£223.39 - £2480.00
Specifications
Accession Number | NP_004765.2 |
---|---|
Gene ID (Entrez) | 5469 |
Name | MED1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16029560
|
enQuireBio™
QP7584-EC-100UG |
100 μg |
£560.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16049560
|
enQuireBio™
QP7584-EC-1MG |
1 mg |
£2480.00
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16039560
|
enQuireBio™
QP7584-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16059560
|
enQuireBio™
QP7584-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069560
|
enQuireBio™
QP7584-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079560
|
enQuireBio™
QP7584-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_004765.2 | |
MED1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
5469 | |
Research Use Only | |
MED1 | |
Recombinant Protein | |
E. coli | |
FGEEYFDESS QSGDNDDFKG FASQALNTLG VPMLGGDNGE TKFKGNNQAD TVDFSIISVA GKALAPADLM EHHSGSQGPL LTTGDLGKEK TQKRVKEGNG TSNSTLSGPG LDSKPGKRSR TPSNDGKSKD KPPKRKKADT EGKSPSHSSS NRPF | |
Greater than 90% as determined by SDS-PAGE. |