missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MED1 Protein

A DNA sequence encoding the Homo sapiens (Human) MED1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7584-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16049560

  • £2480.00 / 1mg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_004765.2
MED1 Protein
Research Use Only
MED1
Recombinant Protein
E. coli
FGEEYFDESS QSGDNDDFKG FASQALNTLG VPMLGGDNGE TKFKGNNQAD TVDFSIISVA GKALAPADLM EHHSGSQGPL LTTGDLGKEK TQKRVKEGNG TSNSTLSGPG LDSKPGKRSR TPSNDGKSKD KPPKRKKADT EGKSPSHSSS NRPF
Greater than 90% as determined by SDS-PAGE.
5469
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers