missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Mast cell carboxypeptidase A Protein
A DNA sequence encoding the Homo sapiens (Human) Mast cell carboxypeptidase A, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001861.2 |
---|---|
Gene ID (Entrez) | 1359 |
Name | Mast cell carboxypeptidase A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15948259
|
enQuireBio™
QP5856-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15968259
|
enQuireBio™
QP5856-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 10-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15958259
|
enQuireBio™
QP5856-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15978259
|
enQuireBio™
QP5856-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15988259
|
enQuireBio™
QP5856-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15998259
|
enQuireBio™
QP5856-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001861.2 | |
Mast cell carboxypeptidase A Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
1359 | |
Research Use Only | |
CPA3 | |
Recombinant Protein | |
E. coli | |
IPGRHSYAKY NNWEKIVAWT EKMMDKYPEM VSRIKIGSTV EDNPLYVLKI GEKNERRKAI FTDCGIHARE WVSPAFCQWF VYQATKTYGR NKIMTKLLDR MNFYILPVFN VDGYIWSWTK NRMWRKNRSK NQNSKCIGTD LNRNFNASWN SIPNTNDPCA DNYRGSAPES EKETKAVTNF IRSHLNEIKV YITFHSYSQM LLFPYGYTSK LPPNHEDLAK VAKIGTDVLS TRYETRYIYG PIESTIYPIS GSSLDWAYDL GIKHTFAFEL RDKGKFGFLL PESRIKPTCR ETMLAVKFIA KYILKHTS | |
Greater than 90% as determined by SDS-PAGE. |