missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Mast cell carboxypeptidase A Protein

A DNA sequence encoding the Homo sapiens (Human) Mast cell carboxypeptidase A, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP5856-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15948259

  • £384.00 / 100µg
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001861.2
Mast cell carboxypeptidase A Protein
Research Use Only
CPA3
Recombinant Protein
E. coli
IPGRHSYAKY NNWEKIVAWT EKMMDKYPEM VSRIKIGSTV EDNPLYVLKI GEKNERRKAI FTDCGIHARE WVSPAFCQWF VYQATKTYGR NKIMTKLLDR MNFYILPVFN VDGYIWSWTK NRMWRKNRSK NQNSKCIGTD LNRNFNASWN SIPNTNDPCA DNYRGSAPES EKETKAVTNF IRSHLNEIKV YITFHSYSQM LLFPYGYTSK LPPNHEDLAK VAKIGTDVLS TRYETRYIYG PIESTIYPIS GSSLDWAYDL GIKHTFAFEL RDKGKFGFLL PESRIKPTCR ETMLAVKFIA KYILKHTS
Greater than 90% as determined by SDS-PAGE.
1359
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers