missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Malate dehydrogenase, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) Malate dehydrogenase, mitochondrial, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001269332.1 |
---|---|
Gene ID (Entrez) | 4191 |
Name | Malate dehydrogenase, mitochondrial Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16062670
|
enQuireBio™
QP6352-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16082670
|
enQuireBio™
QP6352-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 13-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16072670
|
enQuireBio™
QP6352-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16092670
|
enQuireBio™
QP6352-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16002680
|
enQuireBio™
QP6352-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16012680
|
enQuireBio™
QP6352-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001269332.1 | |
Malate dehydrogenase, mitochondrial Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
4191 | |
Research Use Only | |
MDH2 | |
Recombinant Protein | |
E. coli | |
AKVAVLGASG GIGQPLSLLL KNSPLVSRLT LYDIAHTPGV AADLSHIETK AAVKGYLGPE QLPDCLKGCD VVVIPAGVPR KPGMTRDDLF NTNATIVATL TAACAQHCPE AMICVIANPV NSTIPITAEV FKKHGVYNPN KIFGVTTLDI VRANTFVAEL KGLDPARVNV PVIGGHAGKT IIPLISQCTP KVDFPQDQLT ALTGRIQEAG TEVVKAKAGA GSATLSMAYA GARFVFSLVD AMNGKEGVVE CSFVKSQETE CTYFSTPLLL GKKGIEKNLG IGKVSSFEEK MISDAIPELK ASIKKGEDFV KTLK | |
Greater than 90% as determined by SDS-PAGE. |