missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Malate dehydrogenase, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) Malate dehydrogenase, mitochondrial, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6352-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16062670

  • £384.00 / 100µg
Estimated Shipment: 13-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001269332.1
Malate dehydrogenase, mitochondrial Protein
Research Use Only
MDH2
Recombinant Protein
E. coli
AKVAVLGASG GIGQPLSLLL KNSPLVSRLT LYDIAHTPGV AADLSHIETK AAVKGYLGPE QLPDCLKGCD VVVIPAGVPR KPGMTRDDLF NTNATIVATL TAACAQHCPE AMICVIANPV NSTIPITAEV FKKHGVYNPN KIFGVTTLDI VRANTFVAEL KGLDPARVNV PVIGGHAGKT IIPLISQCTP KVDFPQDQLT ALTGRIQEAG TEVVKAKAGA GSATLSMAYA GARFVFSLVD AMNGKEGVVE CSFVKSQETE CTYFSTPLLL GKKGIEKNLG IGKVSSFEEK MISDAIPELK ASIKKGEDFV KTLK
Greater than 90% as determined by SDS-PAGE.
4191
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers