missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-27 beta / EBI3 subunit Protein
Recombinant Protein
£80.80 - £5400.00
Specifications
Accession Number | NP_005746.2 |
---|---|
Gene ID (Entrez) | 10148 |
Name | IL-27 beta / EBI3 subunit Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Less than or equal to 5.00 EUs/ug as determined by the LAL method. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15945849
|
enQuireBio™
QP5284-5UG |
5 μg |
£80.80
5µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15925849
|
enQuireBio™
QP5284-20UG |
20 μg |
£191.00
20µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15905849
|
enQuireBio™
QP5284-100UG |
100 μg |
£715.00
100µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15935849
|
enQuireBio™
QP5284-500UG |
500 μg |
£3285.00
500µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15915849
|
enQuireBio™
QP5284-1MG |
1 mg |
£5400.00
1mg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
NP_005746.2 | |
IL-27 beta / EBI3 subunit Protein | |
Less than or equal to 5.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MRKGPPAALT LPRVQCRASR YPIAVDCSWT LPPAPNSTSP VSFIATYRLG MAARGHSWPC LQQTPTSTSC TITDVQLFSM APYVLNVTAV HPWGSSSSFV PFITEHIIKP DPPEGVRLSP LAERQLQVQW EPPGSWPFPE IFSLKYWIRY KRQGAARFHR VGPIEATSFI LRAVRPRARY YVQVAAQDLT DYGELSDWSL PATATMSLGK | |
Greater than 90 as determined by SDS-PAGE. |
10148 | |
Research Use Only | |
EBI3 | |
Human | |
Untagged | |
0.1% Trifluoroacetic Acid (TFA) + 0.5% mannitol |