Learn More
enQuireBio™ Recombinant Human IL-27 beta / EBI3 subunit Protein
Recombinant Protein
Brand:  enQuireBio™ QP5284-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15905849
  • £715.00 / 100µg
Estimated Shipment: 30-05-2024
to see stock.
Specifications
NP_005746.2
IL-27 beta / EBI3 subunit Protein
Research Use Only
EBI3
Human
Untagged
0.1% Trifluoroacetic Acid (TFA) + 0.5% mannitol
10148
100 μg
Less than or equal to 5.00 EUs/ug as determined by the LAL method.
Recombinant Protein
E. coli
MRKGPPAALT LPRVQCRASR YPIAVDCSWT LPPAPNSTSP VSFIATYRLG MAARGHSWPC LQQTPTSTSC TITDVQLFSM APYVLNVTAV HPWGSSSSFV PFITEHIIKP DPPEGVRLSP LAERQLQVQW EPPGSWPFPE IFSLKYWIRY KRQGAARFHR VGPIEATSFI LRAVRPRARY YVQVAAQDLT DYGELSDWSL PATATMSLGK
Greater than 90 as determined by SDS-PAGE.