missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-27 beta / EBI3 subunit Protein
Recombinant Protein
Brand: enQuireBio™ QP5284-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_005746.2 | |
IL-27 beta / EBI3 subunit Protein | |
Research Use Only | |
EBI3 | |
Human | |
Untagged | |
0.1% Trifluoroacetic Acid (TFA) + 0.5% mannitol |
10148 | |
500 μg | |
Less than or equal to 5.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MRKGPPAALT LPRVQCRASR YPIAVDCSWT LPPAPNSTSP VSFIATYRLG MAARGHSWPC LQQTPTSTSC TITDVQLFSM APYVLNVTAV HPWGSSSSFV PFITEHIIKP DPPEGVRLSP LAERQLQVQW EPPGSWPFPE IFSLKYWIRY KRQGAARFHR VGPIEATSFI LRAVRPRARY YVQVAAQDLT DYGELSDWSL PATATMSLGK | |
Greater than 90 as determined by SDS-PAGE. |