missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Heat shock protein beta-2 Protein
A DNA sequence encoding the Homo sapiens (Human) Heat shock protein beta-2, was expressed in the hosts and tags indicated.
£145.46 - £1735.00
Specifications
Accession Number | NP_001532.1 |
---|---|
Gene ID (Entrez) | 3316 |
Name | Heat shock protein beta-2 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16029290
|
enQuireBio™
QP7544-EC-100UG |
100 μg |
£384.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16049290
|
enQuireBio™
QP7544-EC-1MG |
1 mg |
£1735.00
1mg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16039290
|
enQuireBio™
QP7544-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16059290
|
enQuireBio™
QP7544-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16069290
|
enQuireBio™
QP7544-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16079290
|
enQuireBio™
QP7544-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001532.1 | |
Heat shock protein beta-2 Protein | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |
3316 | |
Research Use Only | |
HSPB2 | |
Recombinant Protein | |
E. coli | |
MSGRSVPHAH PATAEYEFAN PSRLGEQRFG EGLLPEEILT PTLYHGYYVR PRAAPAGEGS RAGASELRLS EGKFQAFLDV SHFTPDEVTV RTVDNLLEVS ARHPQRLDRH GFVSREFCRT YVLPADVDPW RVRAALSHDG ILNLEAPRGG RHLDTEVNEV YISLLPAPPD PEEEEEAAIV EP | |
Greater than 80% as determined by SDS-PAGE. |