missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Heat shock protein beta-2 Protein

A DNA sequence encoding the Homo sapiens (Human) Heat shock protein beta-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7544-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029290

  • £384.00 / 100µg
Estimated Shipment: 07-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001532.1
Heat shock protein beta-2 Protein
Research Use Only
HSPB2
Recombinant Protein
E. coli
MSGRSVPHAH PATAEYEFAN PSRLGEQRFG EGLLPEEILT PTLYHGYYVR PRAAPAGEGS RAGASELRLS EGKFQAFLDV SHFTPDEVTV RTVDNLLEVS ARHPQRLDRH GFVSREFCRT YVLPADVDPW RVRAALSHDG ILNLEAPRGG RHLDTEVNEV YISLLPAPPD PEEEEEAAIV EP
Greater than 80% as determined by SDS-PAGE.
3316
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers