missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human HAND1 Protein
A cDNA sequence encoding the HAND1 was constructed and used to recombinantly synthesize the protein.
£99.15 - £2975.00
Specifications
Name | HAND1 Protein |
---|---|
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Cross Reactivity | Human |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15959368
|
enQuireBio™
QP12100-5UG |
5 μg |
£99.15
5µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15949368
|
enQuireBio™
QP12100-20UG |
20 μg |
£155.00
20µg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15939368
|
enQuireBio™
QP12100-1MG |
1 mg |
£2975.00
1mg |
Estimated Shipment: 31-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HAND1 Protein | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The HAND1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.4M urea and 10% glycerol. |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMNLVGSY AHHHHHHHPH PAHPMLHEPF LFGPASRCHQ ERPYFQSWLL SPADAAPDFP AGGPPPAAAA AATAYGPDAR PGQSPGRLEA LGGRLGRRKG SGPKKERRRT ESINSAFAEL RECIPNVPAD TKLSKIKTLR LATSYIAYLM DVLAKDAQSG DPEAFKAELK KADGGRESKR KRELQQHEGF PPALGPVEKR IKGRTGWPQQ VWALELNQ | |
Greater than 90% as determined by SDS-PAGE. |